Assay Details
Gene Symbol: Atraid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127526.1 | NP_001120998.1 | 2 - 3 | 236 | 92 | |
GenBank mRNA | BC166904.1 | - | 2 - 3 | 236 | 92 | |
FQ214285.1 | - | 2 - 3 | 262 | 92 | ||
FQ222311.1 | - | 2 - 3 | 237 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
