Assay Details
Gene Symbol: Atraid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127526.1 | NP_001120998.1 | 5 - 6 | 488 | 95 | |
GenBank mRNA | BC166904.1 | - | 5 - 6 | 488 | 95 | |
FQ214285.1 | - | 5 - 6 | 514 | 95 | ||
FQ222311.1 | - | 5 - 6 | 489 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
