Assay Details
Gene Symbol: Asxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764542.1 | XP_008762764.1 | 10 - 11 | 1624 | 152 | |
XM_008764543.1 | XP_008762765.1 | 9 - 10 | 1480 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
