Assay Details
Gene Symbol: Eif2s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019356.1 | NP_062229.1 | 2 - 3 | 260 | 101 | |
GenBank mRNA | BC087019.1 | - | 2 - 3 | 317 | 101 | |
J02646.1 | - | 2 - 3 | 260 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
