Assay Details
Gene Symbol: Slc5a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130746.1 | NP_570102.1 | 11 - 12 | 1502 | 60 | |
GenBank mRNA | AF026554.1 | - | 11 - 12 | 1502 | 60 | |
AF081204.2 | - | 8 - 9 | 1090 | 60 | ||
AF081205.1 | - | 8 - 9 | 1090 | 60 | ||
BC168145.1 | - | 9 - 10 | 1364 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
