Assay Details
Gene Symbol: Rad51b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764870.1 | XP_008763092.1 | 2 - 3 | 145 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
