Assay Details
Gene Symbol: Ppp1cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013065.2 | NP_037197.1 | 4 - 5 | 780 | 156 | |
XM_008764505.1 | XP_008762727.1 | 5 - 6 | 718 | 156 | ||
GenBank mRNA | BC062033.1 | - | 4 - 5 | 780 | 156 | |
D90164.1 | - | 4 - 5 | 560 | 156 | ||
FQ212294.1 | - | 5 - 6 | 718 | 156 | ||
S78218.1 | - | 4 - 5 | 547 | 156 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
