Assay Details
Gene Symbol: Ppp1cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764505.1 | XP_008762727.1 | 1 - 2 | 109 | 76 | |
GenBank mRNA | FQ212294.1 | - | 1 - 2 | 109 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008764505.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
