Assay Details
Gene Symbol: Ppp1cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013065.2 | NP_037197.1 | 6 - 7 | 1002 | 219 | |
XM_008764505.1 | XP_008762727.1 | 7 - 8 | 940 | 219 | ||
GenBank mRNA | BC062033.1 | - | 6 - 7 | 1002 | 219 | |
D90164.1 | - | 6 - 7 | 782 | 219 | ||
FQ212294.1 | - | 7 - 8 | 940 | 219 | ||
S78218.1 | - | 6 - 7 | 769 | 219 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
