Assay Details
Gene Symbol: Odc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239906.2 | 11 - 12 | 1608 | 94 | ||
XM_006239907.2 | XP_006239969.1 | 11 - 12 | 1598 | 94 | ||
XM_006239908.2 | XP_006239970.1 | 10 - 11 | 1514 | 94 | ||
XM_008764573.1 | 11 - 12 | 1581 | 94 | |||
GenBank mRNA | BC078882.1 | - | 9 - 10 | 1286 | 94 | |
J04791.1 | - | 11 - 12 | 1402 | 94 | ||
M16982.1 | - | 11 - 12 | 1666 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
