Assay Details
Gene Symbol: Odc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239906.2 | 9 - 10 | 1276 | 172 | ||
XM_006239907.2 | XP_006239969.1 | 9 - 10 | 1266 | 172 | ||
XM_006239908.2 | XP_006239970.1 | 8 - 9 | 1182 | 172 | ||
XM_008764573.1 | 9 - 10 | 1249 | 172 | |||
GenBank mRNA | BC078882.1 | - | 7 - 8 | 954 | 172 | |
J04791.1 | - | 9 - 10 | 1070 | 172 | ||
M16982.1 | - | 9 - 10 | 1334 | 172 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
