Assay Details
Gene Symbol: Hs1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_233975.8 | XP_233975.5 | 1 - 2 | 95 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_233975.8 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
