Assay Details
Gene Symbol: Hs1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_233975.8 | XP_233975.5 | 2 - 3 | 250 | 96 | |
GenBank mRNA | FQ224595.1 | - | 2 - 3 | 219 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
