Assay Details
Gene Symbol: Crnkl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053797.2 | NP_446249.1 | 1 | 1263 | 158 | |
GenBank mRNA | AF245018.1 | - | 1 | 1253 | 158 | |
BC085718.1 | - | 1 | 1263 | 158 |
Gene Symbol: LOC102548514 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235165.2 | XP_006235227.1 | 9 | 1310 | 158 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
