Assay Details
Gene Symbol: Galk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008282.1 | NP_001008283.1 | 6 - 7 | 968 | 67 | |
GenBank mRNA | BC085919.1 | - | 6 - 7 | 968 | 67 | |
FQ219172.1 | - | 6 - 7 | 982 | 67 | ||
FQ234739.1 | - | 6 - 7 | 941 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
