Assay Details
Gene Symbol: Galk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008282.1 | NP_001008283.1 | 7 - 8 | 1140 | 90 | |
GenBank mRNA | BC085919.1 | - | 7 - 8 | 1140 | 90 | |
FQ219172.1 | - | 7 - 8 | 1154 | 90 | ||
FQ234739.1 | - | 7 - 8 | 1113 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
