Assay Details
Gene Symbol: Sept9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113497.1 | NP_001106969.1 | 601 | 102 | ||
NM_031837.2 | NP_114025.2 | 1489 | 102 | |||
NM_176856.2 | NP_789826.2 | 1608 | 102 | |||
GenBank mRNA | AF170253.1 | - | 1609 | 102 | ||
AF173899.1 | - | 1488 | 102 | |||
AF180525.1 | - | 5 | 768 | 102 | ||
AF180526.1 | - | 589 | 102 | |||
BC166602.1 | - | 1154 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
