Assay Details
Gene Symbol: Elp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001718.1 | NP_001001718.1 | 3 - 4 | 453 | 77 | |
XM_006246639.2 | XP_006246701.1 | 4 - 5 | 647 | 77 | ||
GenBank mRNA | AY623900.1 | - | 3 - 4 | 453 | 77 | |
BC158569.1 | - | 4 - 5 | 650 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
