Assay Details
Gene Symbol: Hagh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033349.2 | NP_203500.2 | 4 - 5 | 471 | 94 | |
XM_006245899.1 | XP_006245961.1 | 4 - 5 | 448 | 94 | ||
XM_006245900.1 | XP_006245962.1 | 3 - 4 | 362 | 94 | ||
GenBank mRNA | BC097301.1 | - | 4 - 5 | 423 | 94 | |
FQ229806.1 | - | 4 - 5 | 540 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
