Assay Details
Gene Symbol: Asic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034014.1 | NP_001029186.1 | 3 - 4 | 982 | 145 | |
NM_012892.2 | NP_037024.2 | 3 - 4 | 997 | 145 | ||
GenBank mRNA | U53211.1 | - | 3 - 4 | 982 | 145 | |
Y14635.1 | - | 3 - 4 | 997 | 145 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
