Assay Details
Gene Symbol: Nt5c1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011982.1 | NP_001011982.1 | 2 - 3 | 208 | 73 | |
XM_006239891.2 | XP_006239953.1 | 2 - 3 | 236 | 73 | ||
XM_006239892.2 | XP_006239954.1 | 2 - 3 | 244 | 73 | ||
GenBank mRNA | BC079437.1 | - | 2 - 3 | 208 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
