Assay Details
Gene Symbol: Nt5c1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011982.1 | NP_001011982.1 | 5 - 6 | 1037 | 60 | |
XM_006239888.2 | XP_006239950.1 | 6 - 7 | 1239 | 60 | ||
XM_006239889.2 | XP_006239951.1 | 6 - 7 | 1236 | 60 | ||
XM_006239890.2 | XP_006239952.1 | 6 - 7 | 1116 | 60 | ||
XM_006239891.2 | XP_006239953.1 | 5 - 6 | 1062 | 60 | ||
XM_006239892.2 | XP_006239954.1 | 5 - 6 | 1073 | 60 | ||
GenBank mRNA | BC079437.1 | - | 5 - 6 | 1037 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
