Assay Details
Gene Symbol: Ahr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013149.2 | NP_037281.2 | 10 - 11 | 2427 | 64 | |
GenBank mRNA | AJ821851.1 | - | 10 - 11 | 2400 | 64 | |
AM902286.1 | - | 10 - 11 | 2427 | 64 | ||
U09000.1 | - | 10 - 11 | 2427 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
cell morphogenesis
ovarian follicle development
blood vessel development
branching involved in blood vessel morphogenesis
B cell homeostasis
liver development
blood vessel remodeling
lymphocyte homeostasis
immune system process
negative regulation of systemic arterial blood pressure
cardiac left ventricle morphogenesis
circumferential growth involved in left ventricle morphogenesis
regulation of transcription, DNA-templated
regulation of transcription by RNA polymerase II
transcription by RNA polymerase II
apoptotic process
blood circulation
response to xenobiotic stimulus
response to toxic substance
regulation of gene expression
response to organic cyclic compound
intracellular receptor signaling pathway
prostate gland development
regulation of B cell proliferation
response to estradiol
circadian regulation of gene expression
embryonic hemopoiesis
post-embryonic hemopoiesis
camera-type eye development
T cell homeostasis
positive regulation of cell size
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of RNA polymerase II transcription preinitiation complex assembly
negative regulation of vasoconstriction
positive regulation of growth
positive regulation of transcription by RNA polymerase II
blood vessel morphogenesis
spleen development
reproductive structure development
gland development
smooth muscle tissue development
negative regulation of necrotic cell death
venous blood vessel development
kidney morphogenesis
common bile duct development
cellular response to cAMP
cellular response to glucocorticoid stimulus
glomerulus morphogenesis
blood vessel diameter maintenance
transcription cis-regulatory region binding
DNA-binding transcription factor activity, RNA polymerase II-specific
cis-regulatory region sequence-specific DNA binding
DNA binding
DNA-binding transcription factor activity
nuclear receptor activity
protein binding
transcription factor binding
sequence-specific DNA binding
protein heterodimerization activity
protein dimerization activity
Hsp90 protein binding
E-box binding
negative regulation of transcription by RNA polymerase II
cell morphogenesis
ovarian follicle development
blood vessel development
branching involved in blood vessel morphogenesis
B cell homeostasis
liver development
blood vessel remodeling
lymphocyte homeostasis
immune system process
negative regulation of systemic arterial blood pressure
cardiac left ventricle morphogenesis
circumferential growth involved in left ventricle morphogenesis
regulation of transcription, DNA-templated
regulation of transcription by RNA polymerase II
transcription by RNA polymerase II
apoptotic process
blood circulation
response to xenobiotic stimulus
response to toxic substance
regulation of gene expression
response to organic cyclic compound
intracellular receptor signaling pathway
prostate gland development
regulation of B cell proliferation
response to estradiol
circadian regulation of gene expression
embryonic hemopoiesis
post-embryonic hemopoiesis
camera-type eye development
T cell homeostasis
positive regulation of cell size
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of RNA polymerase II transcription preinitiation complex assembly
negative regulation of vasoconstriction
positive regulation of growth
positive regulation of transcription by RNA polymerase II
blood vessel morphogenesis
spleen development
reproductive structure development
gland development
smooth muscle tissue development
negative regulation of necrotic cell death
venous blood vessel development
kidney morphogenesis
common bile duct development
cellular response to cAMP
cellular response to glucocorticoid stimulus
glomerulus morphogenesis
blood vessel diameter maintenance
transcription cis-regulatory region binding
DNA-binding transcription factor activity, RNA polymerase II-specific
cis-regulatory region sequence-specific DNA binding
DNA binding
DNA-binding transcription factor activity
nuclear receptor activity
protein binding
transcription factor binding
sequence-specific DNA binding
protein heterodimerization activity
protein dimerization activity
Hsp90 protein binding
E-box binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P037843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900016 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119444 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P037843 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P037843 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P037843 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15262 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 43556 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 9285 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10889664 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889673 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889666 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889665 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889668 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889672 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889662 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889663 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889674 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889669 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889667 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889671 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889670 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10889660 | - | - |
Rat Genome 230 2.0 Array | 1369147_at | - | - |
Rat Genome 230 2.0 Array | 1382372_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369147_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369146_a_at | - | - |
Rat Genome 230 2.0 Array | 1369146_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1382372_at | - | - |
Affymetrix HT Rat Focus Array | 1369146_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1369146_a_at | - | - |
Affymetrix HT Rat Focus Array | 1369147_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369147_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0600016784.rn.1 | TC0600002326.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600016780.rn.1 | TC0600002326.rn.1 | RTA TC |
Rat Expression Set 230 | 1369146_a_at | - | - |
Rat Expression Set 230 | 1369147_PM_at | - | - |
Rat Expression Set 230 | 1369147_at | - | - |
Rat Expression Set 230 | 1369146_PM_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10889660 | - | - |
Rat Genome U34 Set | AF082125_s_at | - | - |
Rat Genome U34 Set | AF082124_s_at | - | - |
Rat Genome U34 Set | AF082126_at | - | - |
Rat Genome U34 Set | U04860_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376339 | - | - |
