Assay Details
Gene Symbol: Arhgef33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764441.1 | XP_008762663.1 | 7 - 8 | 935 | 86 | |
XM_008764442.1 | XP_008762664.1 | 7 - 8 | 935 | 86 | ||
XM_008764443.1 | XP_008762665.1 | 7 - 8 | 935 | 86 | ||
XM_008764444.1 | XP_008762666.1 | 7 - 8 | 881 | 86 | ||
XM_008764445.1 | XP_008762667.1 | 7 - 8 | 935 | 86 | ||
XM_008764446.1 | XP_008762668.1 | 7 - 8 | 935 | 86 | ||
XM_008764447.1 | XP_008762669.1 | 7 - 8 | 935 | 86 | ||
XM_008764448.1 | XP_008762670.1 | 7 - 8 | 935 | 86 | ||
XM_008764449.1 | XP_008762671.1 | 7 - 8 | 935 | 86 | ||
XM_008764450.1 | XP_008762672.1 | 7 - 8 | 935 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
