Assay Details
Gene Symbol: Max | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764846.1 | XP_008763068.1 | 1 - 2 | 261 | 89 | |
GenBank mRNA | BC099778.1 | - | 1 - 2 | 192 | 89 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC099778.1, XM_008764846.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
