Assay Details
Gene Symbol: Prkar2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030020.1 | NP_001025191.1 | 8 - 9 | 918 | 139 | |
GenBank mRNA | BC099223.1 | - | 7 - 8 | 661 | 139 | |
M12492.1 | - | 8 - 9 | 912 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
