Assay Details
Gene Symbol: Atp6v1c2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014199.1 | NP_001014221.1 | 4 - 5 | 406 | 68 | |
GenBank mRNA | AY594319.1 | - | 3 - 4 | 331 | 68 | |
BC079083.1 | - | 4 - 5 | 406 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
