Assay Details
Gene Symbol: Timm9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133604.1 | NP_598288.1 | 2 - 3 | 218 | 76 | |
GenBank mRNA | CB775939.1 | - | 2 - 3 | 197 | 76 | |
FQ228524.1 | - | 2 - 3 | 218 | 76 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_133604.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
