Assay Details
Gene Symbol: Timm9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133604.1 | NP_598288.1 | 1 - 2 | 131 | 64 | |
NR_075101.1 | 1 - 2 | 131 | 64 | |||
GenBank mRNA | CB775939.1 | - | 1 - 2 | 110 | 64 | |
DV726219.1 | - | 1 - 2 | 101 | 64 | ||
FQ228524.1 | - | 1 - 2 | 131 | 64 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_133604.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
