Assay Details
Gene Symbol: Iah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100540.1 | NP_001094010.1 | 3 - 4 | 296 | 153 | |
GenBank mRNA | BC168771.1 | - | 3 - 4 | 296 | 153 | |
FQ220941.1 | - | 3 - 4 | 292 | 153 | ||
FQ225724.1 | - | 3 - 4 | 321 | 153 | ||
FQ231316.1 | - | 3 - 4 | 321 | 153 | ||
FQ232084.1 | - | 3 - 4 | 326 | 153 | ||
FQ235324.1 | - | 3 - 4 | 321 | 153 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
