Assay Details
Gene Symbol: Rbks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108703.1 | NP_001102173.1 | 1 - 2 | 125 | 118 | |
XM_006239805.2 | XP_006239867.1 | 1 - 2 | 171 | 118 | ||
XM_006239806.2 | XP_006239868.1 | 1 - 2 | 171 | 118 | ||
XM_008764521.1 | XP_008762743.1 | 1 - 2 | 171 | 118 | ||
GenBank mRNA | FQ209797.1 | - | 1 - 2 | 107 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
