Assay Details
Gene Symbol: Pkdcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108007.1 | NP_001101477.1 | 1 - 2 | 248 | 88 | |
GenBank mRNA | BC166535.1 | - | 1 - 2 | 220 | 88 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC166535.1, NM_001108007.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
