Assay Details
Gene Symbol: Ppm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270619.1 | NP_001257548.1 | 2 - 3 | 1251 | 143 | |
NM_001270620.1 | NP_001257549.1 | 3 - 4 | 1725 | 143 | ||
NM_033096.2 | NP_149087.1 | 2 - 3 | 1251 | 143 | ||
XM_008764454.1 | XP_008762676.1 | 1 - 2 | 862 | 143 | ||
GenBank mRNA | AJ271834.1 | - | 2 - 3 | 1244 | 143 | |
AJ271837.1 | - | 2 - 3 | 1244 | 143 | ||
BC061986.1 | - | 2 - 3 | 1206 | 143 | ||
BC081762.1 | - | 3 - 4 | 1725 | 143 | ||
FM108050.1 | - | 1 - 2 | 406 | 143 | ||
S90449.1 | - | 2 - 3 | 1537 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
