Assay Details
Gene Symbol: Tcea1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025735.1 | NP_001020906.1 | 3 - 4 | 314 | 120 | |
XM_006237816.2 | XP_006237878.1 | 3 - 4 | 375 | 120 | ||
XM_008763546.1 | XP_008761768.1 | 3 - 4 | 202 | 120 | ||
GenBank mRNA | BC099141.1 | - | 3 - 4 | 314 | 120 | |
FQ227501.1 | - | 3 - 4 | 286 | 120 |
Gene Symbol: LOC498453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251720.2 | XP_006251782.1 | 1 | 368 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
