Assay Details
Gene Symbol: Tcea1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025735.1 | NP_001020906.1 | 8 - 9 | 902 | 86 | |
XM_006237816.2 | XP_006237878.1 | 8 - 9 | 963 | 86 | ||
XM_008763546.1 | XP_008761768.1 | 8 - 9 | 790 | 86 | ||
GenBank mRNA | BC099141.1 | - | 8 - 9 | 902 | 86 | |
FQ227501.1 | - | 8 - 9 | 874 | 86 |
Gene Symbol: LOC498453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006251720.2 | XP_006251782.1 | 1 | 956 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription by RNA polymerase II
erythrocyte differentiation
regulation of DNA-templated transcription, elongation
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
positive regulation of exoribonuclease activity
molecular_function
DNA binding
zinc ion binding
transcription, DNA-templated
regulation of transcription by RNA polymerase II
erythrocyte differentiation
regulation of DNA-templated transcription, elongation
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
positive regulation of exoribonuclease activity
molecular_function
DNA binding
zinc ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P084028 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P146124 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P10591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171686 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P146124 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P084028 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P10591 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P146124 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P10591 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P084028 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P084028 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P10591 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P146124 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P897537 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P10591 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6839 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12473 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10875243 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875239 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875234 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875240 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875241 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875238 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875242 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10779354 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875237 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875236 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875244 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875235 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10875233 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10779353 | - | - |
Rat Genome 230 2.0 Array | 1394724_at | - | - |
Rat Genome 230 2.0 Array | 1394724_PM_at | - | - |
Rat Genome 230 2.0 Array | 1382861_PM_at | - | - |
Rat Genome 230 2.0 Array | 1367485_PM_at | - | - |
Rat Genome 230 2.0 Array | 1367485_at | - | - |
Rat Genome 230 2.0 Array | 1392658_PM_at | - | - |
Rat Genome 230 2.0 Array | 1392658_at | - | - |
Rat Genome 230 2.0 Array | 1382861_at | - | - |
Affymetrix HT Rat Focus Array | 1394724_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392658_at | - | - |
Affymetrix HT Rat Focus Array | 1392658_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1367485_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1394724_at | - | - |
Affymetrix HT Rat Focus Array | 1367485_at | - | - |
Affymetrix HT Rat Focus Array | 1382861_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382861_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500016915.rn.1 | TC0500002217.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500016917.rn.1 | TC0500002217.rn.1 | RTA TC |
Rat Expression Set 230 | 1367485_PM_at | - | - |
Rat Expression Set 230 | 1367485_at | - | - |
Rat Expression Set 230 | 1382861_PM_at | - | - |
Rat Expression Set 230 | 1382861_at | - | - |
Rat Expression Set 230 | 1392658_at | - | - |
Rat Expression Set 230 | 1394724_at | - | - |
Rat Expression Set 230 | 1394724_PM_at | - | - |
Rat Expression Set 230 | 1367485_at | - | - |
Rat Expression Set 230 | 1392658_PM_at | - | - |
Rat Expression Set 230 | 1367485_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10779353 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10875233 | - | - |
Rat Genome U34 Set | rc_AA997486_at | - | - |
Rat Genome U34 Set | rc_AI230409_at | - | - |
Rat Genome U34 Set | rc_AI231565_at | - | - |
Rat Genome U34 Set | rc_AI146090_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20864746 | - | - |
ABI Rat Genome Survey Microarray | 20986440 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371058 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359903 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369147 | - | - |
RatRef-12 Expression BeadChip | ILMN_60735 | - | - |
