Assay Details
Gene Symbol: Aip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172327.2 | NP_758830.2 | 5 - 6 | 792 | 85 | |
XM_006230699.2 | XP_006230761.1 | 6 - 7 | 990 | 85 | ||
XM_006230700.2 | XP_006230762.1 | 6 - 7 | 959 | 85 | ||
GenBank mRNA | AF543560.1 | - | 5 - 6 | 826 | 85 | |
BC089110.1 | - | 5 - 6 | 792 | 85 | ||
FQ228146.1 | - | 5 - 6 | 820 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
