Assay Details
Gene Symbol: Pak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053306.3 | NP_445758.2 | 413 | 104 | ||
GenBank mRNA | S80221.1 | - | 332 | 104 | ||
U19967.1 | - | 413 | 104 | |||
U35345.1 | - | 332 | 104 |
Gene Symbol: LOC100910732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751066.2 | XP_003751114.1 | 3 - 4 | 561 | 104 | |
XM_006248473.1 | XP_006248535.1 | 4 - 5 | 690 | 104 | ||
XM_008768778.1 | XP_008767000.1 | 3 - 4 | 2122 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
