Assay Details
Gene Symbol: Poglut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100652.1 | NP_001094122.1 | 3 - 4 | 418 | 149 | |
GenBank mRNA | BC091408.1 | - | 2 - 3 | 190 | 149 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
