Assay Details
Gene Symbol: Tmem39a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013865.1 | NP_001013887.1 | 6 - 7 | 1271 | 80 | |
XM_008768717.1 | XP_008766939.1 | 5 - 6 | 1226 | 80 | ||
GenBank mRNA | BC085845.1 | - | 6 - 7 | 1271 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
