Assay Details
Gene Symbol: Umps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025402.1 | NP_001020573.1 | 3 - 4 | 983 | 124 | |
GenBank mRNA | BC098033.1 | - | 3 - 4 | 983 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
