Assay Details
Gene Symbol: Osbpl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107090.2 | NP_001100560.1 | 3 - 4 | 893 | 56 | |
XM_006248450.2 | XP_006248512.1 | 3 - 4 | 1125 | 56 | ||
XM_006248451.2 | XP_006248513.1 | 3 - 4 | 524 | 56 | ||
XM_006248452.2 | XP_006248514.1 | 3 - 4 | 405 | 56 | ||
XM_008768726.1 | XP_008766948.1 | 3 - 4 | 674 | 56 | ||
GenBank mRNA | BC168952.1 | - | 3 - 4 | 893 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
