Assay Details
Gene Symbol: Slc25a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053501.2 | NP_445953.1 | 2 - 3 | 271 | 65 | |
GenBank mRNA | AF300424.1 | - | 2 - 3 | 272 | 65 | |
AJ300165.1 | - | 1 - 2 | 77 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
