Assay Details
Gene Symbol: Eif3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106242.1 | NP_001099712.1 | 2 - 3 | 218 | 68 | |
XM_006228715.2 | XP_006228777.1 | 5 - 6 | 525 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
