Assay Details
Gene Symbol: Vrk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005561.1 | NP_001005561.1 | 9 - 10 | 900 | 125 | |
XM_006229095.2 | XP_006229157.1 | 9 - 10 | 959 | 125 | ||
XM_006229096.2 | XP_006229158.1 | 9 - 10 | 960 | 125 | ||
GenBank mRNA | BC081928.1 | - | 9 - 10 | 900 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
