Assay Details
Gene Symbol: Gpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207592.1 | NP_997475.1 | 14 - 15 | 1334 | 67 | |
GenBank mRNA | BC062005.1 | - | 14 - 15 | 1334 | 67 | |
FQ215247.1 | - | 3 - 4 | 381 | 67 | ||
FQ216077.1 | - | 3 - 4 | 382 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
