Assay Details
Gene Symbol: Gpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207592.1 | NP_997475.1 | 16 - 17 | 1537 | 67 | |
GenBank mRNA | BC062005.1 | - | 16 - 17 | 1537 | 67 | |
FQ215247.1 | - | 5 - 6 | 584 | 67 | ||
FQ216077.1 | - | 5 - 6 | 585 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
