Assay Details
Gene Symbol: Tgfb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021578.2 | NP_067589.1 | 2 - 3 | 665 | 112 | |
GenBank mRNA | AY550025.1 | - | 2 - 3 | 520 | 112 | |
BC076380.1 | - | 2 - 3 | 665 | 112 | ||
X52498.1 | - | 2 - 3 | 932 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
