Assay Details
Gene Symbol: Fuz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037646.1 | NP_001032735.1 | 9 - 10 | 970 | 97 | |
XM_006229059.2 | XP_006229121.1 | 10 - 11 | 1207 | 97 | ||
XM_006229061.2 | XP_006229123.1 | 10 - 11 | 1260 | 97 | ||
GenBank mRNA | BC107809.1 | - | 9 - 10 | 970 | 97 | |
FQ228777.1 | - | 10 - 11 | 1153 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
