Assay Details
Gene Symbol: Fuz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229059.2 | XP_006229121.1 | 5 - 6 | 665 | 99 | |
XM_006229060.2 | XP_006229122.1 | 5 - 6 | 665 | 99 | ||
XM_006229061.2 | XP_006229123.1 | 5 - 6 | 718 | 99 | ||
XM_008759373.1 | XP_008757595.1 | 5 - 6 | 666 | 99 | ||
XM_008759374.1 | XP_008757596.1 | 5 - 6 | 666 | 99 | ||
GenBank mRNA | FQ228777.1 | - | 5 - 6 | 611 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
