Assay Details
Gene Symbol: Numbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228567.2 | XP_006228629.1 | 1 - 2 | 250 | 123 | |
GenBank mRNA | BC129073.1 | - | 1 - 2 | 137 | 7189989 | 123 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC129073.1, XM_006228567.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
